ContactUsLinkComponent
CCL16 Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 97 amino acids and having a molecular m ass of 11.2 kDa. The CCL16 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80119-20
Supplier:  Biorbyt
CCL16 Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 97 amino acids and having a molecular m ass of 11.2 kDa. The CCL16 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80119-20
Supplier:  Biorbyt