ContactUsLinkComponent
TSLP Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a mo lecular mass of 15 kDa.The TSLP is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80113-10
Supplier:  Biorbyt
TSLP Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a mo lecular mass of 15 kDa.The TSLP is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80113-10
Supplier:  Biorbyt