ContactUsLinkComponent
TNFRSF17 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 50 amino acids and having a molecular mass of 5.3 kDa. The TNFRSF17 is purified by standard chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80112-5
Supplier:  Biorbyt
TNFRSF17 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 50 amino acids and having a molecular mass of 5.3 kDa. The TNFRSF17 is purified by standard chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80112-5
Supplier:  Biorbyt