ContactUsLinkComponent
Sonic HedgeHog Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 175 amino acids and h aving a molecular mass of 19,683 Dalton. The Human Sonic Hedgehog is 99% homologous to the mouse gene.Cystei 1 * 5 µG
  BIRBORB80100-5
 :  Biorbyt
Sonic HedgeHog Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 175 amino acids and h aving a molecular mass of 19,683 Dalton. The Human Sonic Hedgehog is 99% homologous to the mouse gene.Cystei 1 * 5 µG
  BIRBORB80100-5
 :  Biorbyt