ContactUsLinkComponent
sCD40 Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 149 amino acids and having a molecular mass of 16308 Dalton. The sCD40 is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80099-50
Supplier:  Biorbyt
sCD40 Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 149 amino acids and having a molecular mass of 16308 Dalton. The sCD40 is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80099-50
Supplier:  Biorbyt