ContactUsLinkComponent
Otoraplin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids and havin g a molecular mass of 12.7 kDa.The OTOR is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80097-20
Supplier:  Biorbyt
Otoraplin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids and havin g a molecular mass of 12.7 kDa.The OTOR is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80097-20
Supplier:  Biorbyt