Noggin Mouse Recombinant produced in E.coli is non-glycosylated, non-disulfide-linked homodimer consisting of two 206 amino acid pol ypeptide chains, having total molecular mass of approximately 46.2 kDa (each chain 23.1 kDa).Noggin Mouse is 1 * 5 µG
Catalog # BIRBORB80093-5
Supplier: Biorbyt