ContactUsLinkComponent
Keratinocyte Growth Factor-1 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 164 am ino acids and having a molecular mass of 18.9 kDa. The FGF-7 is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB80086-10
Supplier:  Biorbyt
Keratinocyte Growth Factor-1 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 164 am ino acids and having a molecular mass of 18.9 kDa. The FGF-7 is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB80086-10
Supplier:  Biorbyt