ContactUsLinkComponent
Human IL33 Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 160 amino acids and having a m olecular mass of 18 kDa. The IL33 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80083-10
Supplier:  Biorbyt
Human IL33 Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 160 amino acids and having a m olecular mass of 18 kDa. The IL33 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80083-10
Supplier:  Biorbyt