IL20 Human Recombinant produced in E.coli is a single, non-glycosylated homodimeric polypeptide chain containing x 153 amino acids a nd having total molecular mass of 35,212 Dalton. The IL-20 is purified by proprietary chromatographic techni 1 * 10 µG
BIRBORB80079-10
: Biorbyt