ContactUsLinkComponent
IL17A Human Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 264 amino acids (2 chains of 132 aa) and having a molecular mass of 30,250 Dalton. The IL-17 is purified by proprietary chroma 1 * 25 µG
Catalog # BIRBORB80077-25
Supplier:  Biorbyt
IL17A Human Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 264 amino acids (2 chains of 132 aa) and having a molecular mass of 30,250 Dalton. The IL-17 is purified by proprietary chroma 1 * 25 µG
Catalog # BIRBORB80077-25
Supplier:  Biorbyt