ContactUsLinkComponent
IL12 Human Recombinant produced in CHO is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino acids and hav ing molecular weight of 75 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subunits.The IL-1 1 * 2 µG
Catalog # BIRBORB80072-2
Supplier:  Biorbyt
IL12 Human Recombinant produced in CHO is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino acids and hav ing molecular weight of 75 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subunits.The IL-1 1 * 2 µG
Catalog # BIRBORB80072-2
Supplier:  Biorbyt