ContactUsLinkComponent
IL11 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and having a mo lecular mass of 19.2kDa. The Mouse IL-11 is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80071-10
 :  Biorbyt
IL11 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and having a mo lecular mass of 19.2kDa. The Mouse IL-11 is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80071-10
 :  Biorbyt