ContactUsLinkComponent
IL11 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and having a mo lecular mass of 19256.29 Dalton. The IL-11 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80070-2
Supplier:  Biorbyt
IL11 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and having a mo lecular mass of 19256.29 Dalton. The IL-11 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80070-2
Supplier:  Biorbyt