IL-10 Recombinant Rat produced in E.coli is a single, glycosylated polypeptide chain containing 160 amino acids and having a molecul ar mass of 18.7 kDa. The IL10 Mouse is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80069-2
Supplier: Biorbyt