ContactUsLinkComponent
IL-10 Recombinant Mouse produced in E.coli is a single, non-glycosylated polypeptide chain containing 161 amino acids and having a m olecular mass of 18785 Dalton. The IL10 Mouse is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80068-10
Supplier:  Biorbyt
IL-10 Recombinant Mouse produced in E.coli is a single, non-glycosylated polypeptide chain containing 161 amino acids and having a m olecular mass of 18785 Dalton. The IL10 Mouse is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80068-10
Supplier:  Biorbyt