ContactUsLinkComponent
IL3 mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 135 amino acids and having a mol ecular mass of 15100 Dalton. The IL-3 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80058-2
 :  Biorbyt
IL3 mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 135 amino acids and having a mol ecular mass of 15100 Dalton. The IL-3 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80058-2
 :  Biorbyt