Human IL3 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 133 amino acids and having a mol ecular mass of 15000 Dalton. The IL3 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80057-10
Supplier: Biorbyt