ContactUsLinkComponent
Human IL1 beta Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 153 amino acids and having a molecul ar mass of 17000 Dalton.The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80050-2
Supplier:  Biorbyt
Human IL1 beta Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 153 amino acids and having a molecul ar mass of 17000 Dalton.The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80050-2
Supplier:  Biorbyt