ContactUsLinkComponent
Insulin-Like Growth Factor-I Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 ami no acids and having a molecular mass of 7655 Dalton. IGF-I is purified by proprietary chromatographic techni 1 * 100 µG
Catalog # BIRBORB80045-100
Supplier:  Biorbyt
Insulin-Like Growth Factor-I Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 ami no acids and having a molecular mass of 7655 Dalton. IGF-I is purified by proprietary chromatographic techni 1 * 100 µG
Catalog # BIRBORB80045-100
Supplier:  Biorbyt