ContactUsLinkComponent
Interferon-gamma Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 135 amino acids and having a molecular mass of 15609 Dalton.The IFN-gamma is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80043-20
Supplier:  Biorbyt
Interferon-gamma Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 135 amino acids and having a molecular mass of 15609 Dalton.The IFN-gamma is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80043-20
Supplier:  Biorbyt