Interferon-alpha 2b Human Recombinant produced in yeast is a single, glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of approximately 19 kDa.The Interferon-alpha 2b gene was obtained from human leukocy 1 * 20 µG
Catalog # BIRBORB80040-20
Supplier: Biorbyt