ContactUsLinkComponent
Interferon-a 2b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 166 amino acids and having a molecular mass of 19400 Dalton.The Interferon-alpha 2b gene was obtained from human leukocytes.The 1 * 100 µG
Catalog # BIRBORB80039-100
Supplier:  Biorbyt
Interferon-a 2b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 166 amino acids and having a molecular mass of 19400 Dalton.The Interferon-alpha 2b gene was obtained from human leukocytes.The 1 * 100 µG
Catalog # BIRBORB80039-100
Supplier:  Biorbyt