ContactUsLinkComponent
Interferon Alpha Human 2a Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19241 Dalton.The Interferon-a 2a gene was obtained from human leukocytes.The 1 * 100 µG
Catalog # BIRBORB80038-100
Supplier:  Biorbyt
Interferon Alpha Human 2a Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 19241 Dalton.The Interferon-a 2a gene was obtained from human leukocytes.The 1 * 100 µG
Catalog # BIRBORB80038-100
Supplier:  Biorbyt