Granulocyte MCSF Human Recombinant produced in E.coli is a single non-glycosylated polypeptide chain approximately 14.6 kDa, contain ing 128 amino acids. GMCSF was initially characterized as a growth factor that can support the in vitro colo 1 * 5 µG
Catalog # BIRBORB80031-5
Supplier: Biorbyt