ContactUsLinkComponent
Flt3-Ligand Human Recombinant produced in E.coli is non-glycosylated, polypeptide chain containing 155 amino acids and having a mole cular mass of 17605 Dalton. Flt3-Ligand is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80029-10
Supplier:  Biorbyt
Flt3-Ligand Human Recombinant produced in E.coli is non-glycosylated, polypeptide chain containing 155 amino acids and having a mole cular mass of 17605 Dalton. Flt3-Ligand is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80029-10
Supplier:  Biorbyt