Fibroblast Growth Factor -21 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 181 am ino acids and an N-terminal Methionin (bold), having molecular weight of 19.7 kDa (calculated). The amino ac 1 * 5 µG
Catalog # BIRBORB80028-5
Supplier: Biorbyt