ContactUsLinkComponent
FGF 9 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 207 amino acids and having a molecular mass of 23.4 kDa. The FGF-9 is purified by proprietary chromatographic techniques. 1 * 5 µG
  BIRBORB80026-5
 :  Biorbyt
FGF 9 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 207 amino acids and having a molecular mass of 23.4 kDa. The FGF-9 is purified by proprietary chromatographic techniques. 1 * 5 µG
  BIRBORB80026-5
 :  Biorbyt