FGF 2 Human Recombinant (FGF-2) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 155 amino acids and h aving a molecular mass of 17353 Dalton.The FGF-b is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80022-10
Supplier: Biorbyt