ContactUsLinkComponent
FGF acidic Human Recombinant (FGF-1) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141amino acids a nd having a molecular mass of 15967 Dalton.The FGF acidic is purified by proprietary chromatographic techniq 1 * 10 µG
Catalog # BIRBORB80021-10
Supplier:  Biorbyt
FGF acidic Human Recombinant (FGF-1) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141amino acids a nd having a molecular mass of 15967 Dalton.The FGF acidic is purified by proprietary chromatographic techniq 1 * 10 µG
Catalog # BIRBORB80021-10
Supplier:  Biorbyt