ContactUsLinkComponent
Epigen Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 72 amino acids and having a molecular mass of 7.9 kDa. Epigen is purified by proprietary chromatographic techniques. 1 * 25 µG
Catalog # BIRBORB80017-25
Supplier:  Biorbyt
Epigen Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 72 amino acids and having a molecular mass of 7.9 kDa. Epigen is purified by proprietary chromatographic techniques. 1 * 25 µG
Catalog # BIRBORB80017-25
Supplier:  Biorbyt