ContactUsLinkComponent
EMAP-II Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 166 amino acids and having a molecular mass of 18.3 kDa. The EMAP-II is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80016-20
Supplier:  Biorbyt
EMAP-II Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 166 amino acids and having a molecular mass of 18.3 kDa. The EMAP-II is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80016-20
Supplier:  Biorbyt