ContactUsLinkComponent
CDKN2A Human Recombinant produced in E.coli, it's single non-glycosylated polypeptide chain containing 156 amino acids, approximatel y 16.5 kDa.CDKN2A is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80172-10
Supplier:  Biorbyt
CDKN2A Human Recombinant produced in E.coli, it's single non-glycosylated polypeptide chain containing 156 amino acids, approximatel y 16.5 kDa.CDKN2A is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80172-10
Supplier:  Biorbyt