ContactUsLinkComponent
CNTF Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a mo lecular mass of 22706 Dalton. The CNTF is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80012-5
Supplier:  Biorbyt
CNTF Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a mo lecular mass of 22706 Dalton. The CNTF is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80012-5
Supplier:  Biorbyt