ContactUsLinkComponent
BMP2 Human Recombinant produced in E.coli is homodimeric, non-glycosylated, Polypeptide chain containing 115 amino acids and having a molecular mass of 26018 Dalton. The BMP-2 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80007-2
 :  Biorbyt
BMP2 Human Recombinant produced in E.coli is homodimeric, non-glycosylated, Polypeptide chain containing 115 amino acids and having a molecular mass of 26018 Dalton. The BMP-2 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80007-2
 :  Biorbyt