ContactUsLinkComponent
Beta Defensin-3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 45 amino acids and h aving a molecular mass of 5161.2 Dalton. The BD-3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80003-5
Supplier:  Biorbyt
Beta Defensin-3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 45 amino acids and h aving a molecular mass of 5161.2 Dalton. The BD-3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80003-5
Supplier:  Biorbyt