ContactUsLinkComponent
TECK Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids and having a m olecular mass of 14.2kDa. The TECK is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80165-5
Supplier:  Biorbyt
TECK Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 127 amino acids and having a m olecular mass of 14.2kDa. The TECK is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80165-5
Supplier:  Biorbyt