ContactUsLinkComponent
CCL28 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids and having a molecular mass of 12.6 kDa. The CCL28 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80148-20
Supplier:  Biorbyt
CCL28 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids and having a molecular mass of 12.6 kDa. The CCL28 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80148-20
Supplier:  Biorbyt