ContactUsLinkComponent
CCL28 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 108 amino acids and having a molecular mass of 12.3 kDa. The CCL28 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80147-20
Supplier:  Biorbyt
CCL28 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 108 amino acids and having a molecular mass of 12.3 kDa. The CCL28 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80147-20
Supplier:  Biorbyt