MDC Human Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 69 amino acids and having a molecular mas s of 8.1 kDa. The MDC is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80145-20
Supplier: Biorbyt