ContactUsLinkComponent
Human ENA78/CXCL5 ELISA Kit for Serum, Plasma, Cell Culture Supernatants and Urine. 1 * 96 items
Catalog # BIRBORB78748-96
Supplier:  Biorbyt
Human ENA78/CXCL5 ELISA Kit for Serum, Plasma, Cell Culture Supernatants and Urine. 1 * 96 items
Catalog # BIRBORB78748-96
Supplier:  Biorbyt