ContactUsLinkComponent
This assay is a two-site ELISA. The micro-plate is pre-coated with a monoclonal antibody against insulin. Standards and samples are added into the wells and co-incubated with a monoclonal antibody conjugated to horseradish peroxidase (HRP) 1 * 96 items
Catalog # BIRBORB54822-96
Supplier:  Biorbyt
This assay is a two-site ELISA. The micro-plate is pre-coated with a monoclonal antibody against insulin. Standards and samples are added into the wells and co-incubated with a monoclonal antibody conjugated to horseradish peroxidase (HRP) 1 * 96 items
Catalog # BIRBORB54822-96
Supplier:  Biorbyt