ContactUsLinkComponent
Beta defensin 3, also known as BD3 and DEFB3, is a membrane active cationic peptide that functions in inflammation and innate immune responses and coded by Defb 3 gene on chromosome 8 in mouse. There are at least 30 beta-defensins which are 1 * 20 g
Catalog # BIRBORB82606-20
Supplier:  Biorbyt
Beta defensin 3, also known as BD3 and DEFB3, is a membrane active cationic peptide that functions in inflammation and innate immune responses and coded by Defb 3 gene on chromosome 8 in mouse. There are at least 30 beta-defensins which are 1 * 20 g
Catalog # BIRBORB82606-20
Supplier:  Biorbyt