ContactUsLinkComponent
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain A/Shandong/9/93. The Influenza Virus was pu rified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81799-50
Supplier:  Biorbyt
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain A/Shandong/9/93. The Influenza Virus was pu rified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81799-50
Supplier:  Biorbyt