ContactUsLinkComponent
HCV-NS3-S6 (residues 1359-1456) protein.The protein contains the HCV NS3 (c33c) immunodominant region, amino acids 1359-1456.Hepatit is C NS3 protein is purified by proprietarychromatographic techniques.The nonstructural protein NS3 of the h 1 * 1 mg
Catalog # BIRBORB97932-1
Supplier:  Biorbyt
HCV-NS3-S6 (residues 1359-1456) protein.The protein contains the HCV NS3 (c33c) immunodominant region, amino acids 1359-1456.Hepatit is C NS3 protein is purified by proprietarychromatographic techniques.The nonstructural protein NS3 of the h 1 * 1 mg
Catalog # BIRBORB97932-1
Supplier:  Biorbyt