ContactUsLinkComponent
HCV-NS3-S2b (residues 1192-1459) protein.The protein contains the full-length HCV NS3 (c33c)immunodominant region, amino acids 1192- 1459.Hepatitis C NS3 protein is purified by proprietarychromatographic techniques.The nonstructural protein 1 * 1 mg
Catalog # BIRBORB97915-1
Supplier:  Biorbyt
HCV-NS3-S2b (residues 1192-1459) protein.The protein contains the full-length HCV NS3 (c33c)immunodominant region, amino acids 1192- 1459.Hepatitis C NS3 protein is purified by proprietarychromatographic techniques.The nonstructural protein 1 * 1 mg
Catalog # BIRBORB97915-1
Supplier:  Biorbyt