ContactUsLinkComponent
Human FGF 9 ELISA Kit for Serum, Plasma, Cell Culture Supernatant and Urine. 1 * 96 items
Catalog # BIRBORB78767-96
Supplier:  Biorbyt
Human FGF 9 ELISA Kit for Serum, Plasma, Cell Culture Supernatant and Urine. 1 * 96 items
Catalog # BIRBORB78767-96
Supplier:  Biorbyt