ContactUsLinkComponent
PM/Scl 100 Human Recombinant produced in SF9 is glycosylated, polypeptide chain having a molecular mass of 120 kDa. PM/Scl 100 is ex pressed with -6xHis tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81278-5
Supplier:  Biorbyt
PM/Scl 100 Human Recombinant produced in SF9 is glycosylated, polypeptide chain having a molecular mass of 120 kDa. PM/Scl 100 is ex pressed with -6xHis tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81278-5
Supplier:  Biorbyt