ContactUsLinkComponent
PCNA Human Recombinant produced in SF9 is glycosylated, polypeptide chain having a molecular mass of 29,794 Dalton. PCNA is expresse d with -6xHis tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81260-5
Supplier:  Biorbyt
PCNA Human Recombinant produced in SF9 is glycosylated, polypeptide chain having a molecular mass of 29,794 Dalton. PCNA is expresse d with -6xHis tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81260-5
Supplier:  Biorbyt