ContactUsLinkComponent
PCNA Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 261 amino acids and having a mo lecular mass of 28769 Dalton. 1 * 5 µG
Catalog # BIRBORB81258-5
Supplier:  Biorbyt
PCNA Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 261 amino acids and having a mo lecular mass of 28769 Dalton. 1 * 5 µG
Catalog # BIRBORB81258-5
Supplier:  Biorbyt